Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAGGAGTGATATTCTAAGGGTTA[A/G]TGACCAAGAGCTTTCCTGAAAAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603869 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAB27B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAB27B - RAB27B, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004163.4 | Intron | NP_004154.2 | ||||
XM_005266733.1 | Intron | XP_005266790.1 | ||||
XM_006722518.1 | Intron | XP_006722581.1 | ||||
XM_011526131.1 | Intron | XP_011524433.1 | ||||
XM_017025913.1 | Intron | XP_016881402.1 | ||||
XM_017025914.1 | Intron | XP_016881403.1 |