Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGTGAGCGATATAGGAATATTAA[C/T]TTGGCATAGAGATCTTCTTCTAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613629 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101927410 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101927410 - uncharacterized LOC101927410 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIEZO2 - piezo type mechanosensitive ion channel component 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022068.3 | 9300 | Silent Mutation | AAA,AAG | K,K 2732 | NP_071351.2 | |
XM_011525723.2 | 9300 | Silent Mutation | AAA,AAG | K,K 2776 | XP_011524025.1 | |
XM_011525724.2 | 9300 | Silent Mutation | AAA,AAG | K,K 2757 | XP_011524026.1 | |
XM_011525725.2 | 9300 | Silent Mutation | AAA,AAG | K,K 2746 | XP_011524027.1 | |
XM_011525726.2 | 9300 | Silent Mutation | AAA,AAG | K,K 2715 | XP_011524028.1 | |
XM_017025918.1 | 9300 | Silent Mutation | AAA,AAG | K,K 2763 | XP_016881407.1 |