Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGCTGCAGATGTTAAGCAGTTG[C/G]GGGGCTCTGTGGAACTGGTGGATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 169800 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNDP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNDP2 - CNDP dipeptidase 2 (metallopeptidase M20 family) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168499.1 | 258 | Missense Mutation | CGG,GGG | R,G 56 | NP_001161971.1 | |
NM_018235.2 | 258 | Missense Mutation | CGG,GGG | R,G 56 | NP_060705.2 | |
XM_005266728.2 | 258 | Missense Mutation | CGG,GGG | R,G 56 | XP_005266785.1 | |
XM_006722503.2 | 258 | Missense Mutation | CGG,GGG | R,G 56 | XP_006722566.1 | |
XM_011526071.2 | 258 | Missense Mutation | CGG,GGG | R,G 56 | XP_011524373.1 | |
XM_011526072.2 | 258 | Missense Mutation | CGG,GGG | R,G 56 | XP_011524374.1 | |
XM_011526073.2 | 258 | UTR 5 | XP_011524375.1 |