Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTAAACATGTAAACAAATCACAGA[C/T]ACTTTAAAAAGTCCAGATTCAGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
L3MBTL4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
L3MBTL4 - l(3)mbt-like 4 (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173464.3 | 2292 | UTR 3 | NP_775735.2 | |||
XM_005258166.1 | 2292 | UTR 3 | XP_005258223.1 | |||
XM_006722364.2 | 2292 | Intron | XP_006722427.1 | |||
XM_011525757.1 | 2292 | UTR 3 | XP_011524059.1 | |||
XM_011525758.1 | 2292 | UTR 3 | XP_011524060.1 | |||
XM_011525759.1 | 2292 | UTR 3 | XP_011524061.1 | |||
XM_011525760.1 | 2292 | UTR 3 | XP_011524062.1 | |||
XM_011525761.1 | 2292 | UTR 3 | XP_011524063.1 | |||
XM_011525762.1 | 2292 | Intron | XP_011524064.1 | |||
XM_011525763.2 | 2292 | Intron | XP_011524065.1 | |||
XM_011525765.2 | 2292 | Intron | XP_011524067.1 | |||
XM_011525767.2 | 2292 | UTR 3 | XP_011524069.1 | |||
XM_017026074.1 | 2292 | UTR 3 | XP_016881563.1 | |||
XM_017026075.1 | 2292 | UTR 3 | XP_016881564.1 | |||
XM_017026076.1 | 2292 | UTR 3 | XP_016881565.1 | |||
XM_017026077.1 | 2292 | Intron | XP_016881566.1 |