Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAAAAATGAGTCTTTCACCTACT[A/G]TTGTTTGCATTGAGAATCCATCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611402 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DOK6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DOK6 - docking protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152721.5 | Intron | NP_689934.2 | ||||
XM_011525875.2 | Intron | XP_011524177.1 | ||||
XM_017025610.1 | Intron | XP_016881099.1 | ||||
XM_017025611.1 | Intron | XP_016881100.1 |