Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTCCTTAAAAATACTTTATCTT[A/C]AGCCATTTTGGTCATAGTCTTTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610974 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF521 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF521 - zinc finger protein 521 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308225.1 | 4455 | UTR 3 | NP_001295154.1 | |||
NM_015461.2 | 4455 | UTR 3 | NP_056276.1 | |||
XM_011525909.2 | 4455 | UTR 3 | XP_011524211.1 | |||
XM_011525910.2 | 4455 | Intron | XP_011524212.1 | |||
XM_011525911.1 | 4455 | UTR 3 | XP_011524213.1 | |||
XM_017025697.1 | 4455 | UTR 3 | XP_016881186.1 | |||
XM_017025698.1 | 4455 | UTR 3 | XP_016881187.1 |