Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGACCATACAGTGCAGCTTCAGGA[C/G]CTGTCCATTTGATTGGAAATTTTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 164880 | ||||||||||||||||||||
Literature Links: |
YES1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
YES1 - YES proto-oncogene 1, Src family tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005433.3 | 1540 | Missense Mutation | GCT,GGT | A,G 440 | NP_005424.1 | |
XM_005258139.3 | 1540 | Intron | XP_005258196.1 | |||
XM_017025960.1 | 1540 | Missense Mutation | GCT,GGT | A,G 440 | XP_016881449.1 |