Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCGGGCGAGGACCCCAATGGTA[C/T]GCTGATCATCCAGAATGTGAACAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601855 MIM: 112205 MIM: 603474 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGEF1 - Rho guanine nucleotide exchange factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CD79A - CD79a molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001783.3 | 454 | Missense Mutation | ACG,ATG | T,M 90 | NP_001774.1 | |
NM_021601.3 | 454 | Intron | NP_067612.1 |
MIR6797 - microRNA 6797 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPS19 - ribosomal protein S19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |