Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGCCTGGTGATGGGATGCATCA[C/T]GGTACTGCTTGGCTGTGCCGGGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSPC102 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HSPC102 - uncharacterized HSPC102 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSPAN16 - tetraspanin 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282509.1 | 293 | Missense Mutation | ACG,ATG | T,M 68 | NP_001269438.1 | |
NM_001282510.1 | 293 | Missense Mutation | ACG,ATG | T,M 68 | NP_001269439.1 | |
NM_012466.3 | 293 | Missense Mutation | ACG,ATG | T,M 68 | NP_036598.1 | |
XM_011527901.1 | 293 | Missense Mutation | ACG,ATG | T,M 68 | XP_011526203.1 | |
XM_011527902.1 | 293 | Missense Mutation | ACG,ATG | T,M 68 | XP_011526204.1 | |
XM_011527903.2 | 293 | Missense Mutation | ACG,ATG | T,M 5 | XP_011526205.1 |