Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGAAGCAATGCTGATGCTCCGGAT[A/G]GTGTCAGACTCGGACACGGATTCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603767 | ||||||||||||||||||||
Literature Links: |
KLK4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KLK4 - kallikrein related peptidase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302961.1 | 393 | Silent Mutation | ACC,ACT | T,T 36 | NP_001289890.1 | |
NM_004917.4 | 393 | Silent Mutation | ACC,ACT | T,T 131 | NP_004908.4 | |
XM_005259441.4 | 393 | Silent Mutation | ACC,ACT | T,T 36 | XP_005259498.2 | |
XM_011527545.2 | 393 | Silent Mutation | ACC,ACT | T,T 131 | XP_011525847.1 | |
XM_011527546.2 | 393 | Silent Mutation | ACC,ACT | T,T 131 | XP_011525848.1 |