Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGAGCATTTACTCTGCGCCAGTC[A/G]TGGTAAGTACTGTACTTCACATGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614591 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEACAM16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEACAM16 - carcinoembryonic antigen related cell adhesion molecule 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039213.3 | 108 | UTR 5 | NP_001034302.2 | |||
XM_017026795.1 | 108 | Intron | XP_016882284.1 |