Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGGATCCACTTACCAGAGACTT[C/T]GACTGTCATGGATTTGGAGCTTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176390 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PSG1 - pregnancy specific beta-1-glycoprotein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184825.1 | 1372 | Missense Mutation | AAA,GAA | K,E 412 | NP_001171754.1 | |
NM_001184826.1 | 1372 | Missense Mutation | AAA,GAA | K,E 412 | NP_001171755.1 | |
NM_001297773.1 | 1372 | Missense Mutation | AAA,GAA | K,E 412 | NP_001284702.1 | |
NM_006905.2 | 1372 | Missense Mutation | AAA,GAA | K,E 412 | NP_008836.2 | |
XM_005259065.3 | 1372 | Missense Mutation | AAA,GAA | K,E 319 | XP_005259122.1 | |
XM_011527125.1 | 1372 | Missense Mutation | AAA,GAA | K,E 319 | XP_011525427.1 | |
XM_017026994.1 | 1372 | Missense Mutation | AAA,GAA | K,E 319 | XP_016882483.1 |