Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTACGGCCAACCTGGGACCCTACTT[C/T]GAGGGGCTCCGCAAGCACTACCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610622 MIM: 610197 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FUZ PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FUZ - fuzzy planar cell polarity protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MED25 - mediator complex subunit 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030973.3 | 159 | Silent Mutation | TTC,TTT | F,F 32 | NP_112235.2 | |
XM_011527353.1 | 159 | Silent Mutation | TTC,TTT | F,F 32 | XP_011525655.1 |