Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGGCGGCCAGGCGGTGGGCCAGC[A/G]GGCCGTCAGGCGGCGGAGGCGACGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609932 MIM: 604125 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM83E PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM83E - family with sequence similarity 83 member E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017708.3 | 1057 | Missense Mutation | CCG,CTG | P,L 331 | NP_060178.2 | |
XM_005259019.1 | 1057 | Missense Mutation | CCG,CTG | P,L 331 | XP_005259076.1 | |
XM_011527049.2 | 1057 | Intron | XP_011525351.1 |
SPACA4 - sperm acrosome associated 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SULT2B1 - sulfotransferase family 2B member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |