Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCAGAGGCCACTCCGGGCTCATC[A/G]AGACCACCGGCCGTGCCATCCACGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929060 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101929060 - uncharacterized LOC101929060 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIER2 - MIER family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005259582.4 | 1826 | Silent Mutation | XP_005259639.1 | |||
XM_006722769.3 | 1826 | Silent Mutation | XP_006722832.1 | |||
XM_011528072.2 | 1826 | Silent Mutation | XP_011526374.1 | |||
XM_011528073.1 | 1826 | Silent Mutation | XP_011526375.1 | |||
XM_011528074.2 | 1826 | Silent Mutation | XP_011526376.1 | |||
XM_011528075.2 | 1826 | Silent Mutation | XP_011526377.1 | |||
XM_011528077.2 | 1826 | Silent Mutation | XP_011526379.1 | |||
XM_011528081.2 | 1826 | Silent Mutation | XP_011526383.1 | |||
XM_011528082.2 | 1826 | Silent Mutation | XP_011526384.1 |