Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGGCTGGCGGCTGAAGTGGCCAC[A/G]AGACTGTGTCCCCAGGAGTGAGTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609694 MIM: 605110 | ||||||||||||||||||||
Literature Links: |
GMIP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GMIP - GEM interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288998.1 | 2635 | Missense Mutation | CGT,TGT | R,C 833 | NP_001275927.1 | |
NM_001288999.1 | 2635 | Missense Mutation | CGT,TGT | R,C 836 | NP_001275928.1 | |
NM_016573.3 | 2635 | Missense Mutation | CGT,TGT | R,C 862 | NP_057657.2 | |
XM_005259927.2 | 2635 | Missense Mutation | CGT,TGT | R,C 859 | XP_005259984.1 | |
XM_011528048.2 | 2635 | Missense Mutation | CGT,TGT | R,C 741 | XP_011526350.1 | |
XM_017026861.1 | 2635 | Silent Mutation | CTC,CTT | L,L 876 | XP_016882350.1 | |
XM_017026862.1 | 2635 | Intron | XP_016882351.1 |
LPAR2 - lysophosphatidic acid receptor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |