Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCCCATCATGCAGGGCCATGTGA[A/C]CTTTGAGGATATTGCTGTGTACTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF549 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF549 - zinc finger protein 549 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199295.1 | 407 | Missense Mutation | AAC,ACC | N,T 28 | NP_001186224.1 | |
NM_153263.2 | 407 | Missense Mutation | AAC,ACC | N,T 15 | NP_694995.2 | |
XM_006723128.3 | 407 | Missense Mutation | AAC,ACC | N,T 6 | XP_006723191.1 |
ZNF550 - zinc finger protein 550 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |