Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAGGGTACCAGGAATAAGTTGAC[C/T]TCAGAGTCCAGGAAGTCAGGTGGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613176 | ||||||||||||||||||||
Literature Links: |
SMG9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SMG9 - SMG9, nonsense mediated mRNA decay factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005259057.2 | 1379 | Silent Mutation | GAA,GAG | E,E 425 | XP_005259114.1 | |
XM_011527113.1 | 1379 | Silent Mutation | GAA,GAG | E,E 425 | XP_011525415.1 | |
XM_011527114.1 | 1379 | Silent Mutation | GAA,GAG | E,E 425 | XP_011525416.1 | |
XM_011527115.1 | 1379 | Silent Mutation | GAA,GAG | E,E 425 | XP_011525417.1 | |
XM_011527116.1 | 1379 | Silent Mutation | GAA,GAG | E,E 425 | XP_011525418.1 | |
XM_017026988.1 | 1379 | Silent Mutation | GAA,GAG | E,E 181 | XP_016882477.1 |