Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATGTCTTTCAAGGTGTGATTTGC[G/T]AATGTAAACTTTGTCACATTCTTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606427 | ||||||||||||||||||||
Literature Links: |
ZNF320 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF320 - zinc finger protein 320 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207333.2 | 1599 | Missense Mutation | NP_997216.2 | |||
XM_006723059.3 | 1599 | Missense Mutation | AGC,CGC | S,R 424 | XP_006723122.1 | |
XM_017026393.1 | 1599 | Missense Mutation | AGC,CGC | S,R 424 | XP_016881882.1 | |
XM_017026394.1 | 1599 | Missense Mutation | AGC,CGC | S,R 424 | XP_016881883.1 | |
XM_017026395.1 | 1599 | Missense Mutation | AGC,CGC | S,R 424 | XP_016881884.1 | |
XM_017026396.1 | 1599 | Missense Mutation | AGC,CGC | S,R 424 | XP_016881885.1 | |
XM_017026397.1 | 1599 | Missense Mutation | AGC,CGC | S,R 424 | XP_016881886.1 | |
XM_017026398.1 | 1599 | Intron | XP_016881887.1 | |||
XM_017026399.1 | 1599 | Missense Mutation | AGC,CGC | S,R 424 | XP_016881888.1 | |
XM_017026400.1 | 1599 | Missense Mutation | AGC,CGC | S,R 424 | XP_016881889.1 |