Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCAGCTCCCAGTGACCCTCTGGAC[A/G]TCGTGATCATAGGTGAGAGTGTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604936 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIR2DL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIR2DL1 - killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014218.2 | 1001 | Missense Mutation | ATC,GTC | I,V 120 | NP_055033.2 | |
XM_011526939.2 | 1001 | Intron | XP_011525241.1 | |||
XM_017026782.1 | 1001 | Missense Mutation | ATC,GTC | I,V 67 | XP_016882271.1 | |
XM_017026783.1 | 1001 | Missense Mutation | ATC,GTC | I,V 120 | XP_016882272.1 |
LOC101928804 - uncharacterized LOC101928804 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |