Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAGAGCCAGGAAGAGGTAGAGGT[A/G]GCAGGAATTAAACTTTGTAAAGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF470 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF470 - zinc finger protein 470 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001668.3 | 703 | Silent Mutation | GTA,GTG | V,V 9 | NP_001001668.3 | |
XM_017026801.1 | 703 | UTR 5 | XP_016882290.1 |