Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614143 MIM: 606834 MIM: 607632 MIM: 601080 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGFLR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IGFLR1 - IGF like family receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024660.2 | 903 | Missense Mutation | CCT,GCT | P,A 269 | NP_078936.1 |
KMT2B - lysine methyltransferase 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014727.2 | 903 | Intron | NP_055542.1 | |||
XM_011527561.2 | 903 | Intron | XP_011525863.2 | |||
XM_011527562.2 | 903 | Intron | XP_011525864.1 | |||
XM_017027544.1 | 903 | Intron | XP_016883033.1 | |||
XM_017027545.1 | 903 | Intron | XP_016883034.1 | |||
XM_017027546.1 | 903 | Intron | XP_016883035.1 |
LIN37 - lin-37 DREAM MuvB core complex component | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSENEN - presenilin enhancer gamma-secretase subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
U2AF1L4 - U2 small nuclear RNA auxiliary factor 1 like 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |