Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCCTCCAGCACAGCCATGTCCCA[C/T]GAGAAGGTCAGGAGGGCAGAGGGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601246 MIM: 611946 | ||||||||||||||||||||
Literature Links: |
CHAF1A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHAF1A - chromatin assembly factor 1 subunit A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005483.2 | 1755 | Intron | NP_005474.2 | |||
XM_011527605.2 | 1755 | Intron | XP_011525907.1 | |||
XM_011527606.2 | 1755 | Intron | XP_011525908.1 | |||
XM_011527607.2 | 1755 | Intron | XP_011525909.1 |
MIR4746 - microRNA 4746 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBXN6 - UBX domain protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171091.1 | 1755 | Silent Mutation | NP_001164562.1 | |||
NM_025241.2 | 1755 | Silent Mutation | NP_079517.1 | |||
XM_017027325.1 | 1755 | Silent Mutation | XP_016882814.1 |