Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGTAAATGATGGCGGATGCCAAG[C/T]ATGTCCTCTGCCGATGGGAAAAGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
MUM1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MUM1 - melanoma associated antigen (mutated) 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032853.3 | 119 | Missense Mutation | CAT,TAT | H,Y 7 | NP_116242.2 | |
XM_011528380.2 | 119 | Missense Mutation | CAT,TAT | H,Y 7 | XP_011526682.1 | |
XM_011528382.2 | 119 | Missense Mutation | CAT,TAT | H,Y 7 | XP_011526684.1 | |
XM_011528383.1 | 119 | Missense Mutation | CAT,TAT | H,Y 7 | XP_011526685.1 | |
XM_011528384.2 | 119 | Missense Mutation | CAT,TAT | H,Y 7 | XP_011526686.1 | |
XM_011528385.2 | 119 | UTR 5 | XP_011526687.1 | |||
XM_017027401.1 | 119 | Missense Mutation | CAT,TAT | H,Y 7 | XP_016882890.1 |