Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAGGCAACTACACCTGCCTGTTC[A/G]TCACGTTCCCGCAGGGCAGCAGGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 173850 | ||||||||||||||||||||
Literature Links: |
MIR4531 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR4531 - microRNA 4531 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PVR - poliovirus receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135768.2 | 675 | Missense Mutation | ATC,GTC | I,V 126 | NP_001129240.1 | |
NM_001135769.2 | 675 | Missense Mutation | ATC,GTC | I,V 126 | NP_001129241.1 | |
NM_001135770.3 | 675 | Missense Mutation | ATC,GTC | I,V 126 | NP_001129242.2 | |
NM_006505.4 | 675 | Missense Mutation | ATC,GTC | I,V 126 | NP_006496.4 |