Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTAGTGCTTATCCTAGAATCAGCC[C/G]TTGCTGCCTATGATCTTCTTGATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605644 MIM: 605504 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLK8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLK8 - kallikrein related peptidase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281431.1 | 533 | Missense Mutation | AAC,AAG | N,K 138 | NP_001268360.1 | |
NM_007196.3 | 533 | Missense Mutation | AAC,AAG | N,K 259 | NP_009127.1 | |
NM_144505.2 | 533 | Missense Mutation | AAC,AAG | N,K 304 | NP_653088.1 | |
NM_144506.2 | 533 | Missense Mutation | AAC,AAG | N,K 118 | NP_653089.1 | |
NM_144507.2 | 533 | UTR 3 | NP_653090.1 |
KLK9 - kallikrein related peptidase 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |