Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGTCATCATTGATTTGACAGAG[G/T]ACTCGAATGAGCAGCCAGACAGTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601246 MIM: 601768 | ||||||||||||||||||||
Literature Links: |
CHAF1A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHAF1A - chromatin assembly factor 1 subunit A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005483.2 | 464 | Missense Mutation | GAC,TAC | D,Y 122 | NP_005474.2 | |
XM_011527605.2 | 464 | Missense Mutation | GAC,TAC | D,Y 122 | XP_011525907.1 | |
XM_011527606.2 | 464 | Missense Mutation | GAC,TAC | D,Y 122 | XP_011525908.1 | |
XM_011527607.2 | 464 | Missense Mutation | GAC,TAC | D,Y 122 | XP_011525909.1 |
SH3GL1 - SH3 domain containing GRB2 like 1, endophilin A2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |