Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGGTCTTGGCGCCCGTGGTGGG[C/T]GACTCGTGTGTCACCTTGCGCAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608531 MIM: 602214 | ||||||||||||||||||||
Literature Links: |
BTBD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BTBD2 - BTB domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017797.3 | 836 | Silent Mutation | TCA,TCG | S,S 490 | NP_060267.2 | |
XM_005259593.1 | 836 | Silent Mutation | TCA,TCG | S,S 342 | XP_005259650.1 | |
XM_011528127.1 | 836 | Silent Mutation | TCA,TCG | S,S 169 | XP_011526429.1 |
CSNK1G2 - casein kinase 1 gamma 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |