Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCGCCTGCCAGTCGCAGCAGGGC[C/T]AGGGTCAGGTGTAGGTTCTGAGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609171 MIM: 616575 | ||||||||||||||||||||
Literature Links: |
CDC42EP5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDC42EP5 - CDC42 effector protein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LENG8 - leukocyte receptor cluster member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052925.3 | 1214 | Intron | NP_443157.1 | |||
XM_005278248.4 | 1214 | Intron | XP_005278305.1 | |||
XM_005278249.4 | 1214 | Intron | XP_005278306.1 | |||
XM_005278250.4 | 1214 | Intron | XP_005278307.1 | |||
XM_005278252.4 | 1214 | Intron | XP_005278309.1 | |||
XM_006722996.3 | 1214 | Intron | XP_006723059.1 | |||
XM_006722997.3 | 1214 | Intron | XP_006723060.1 | |||
XM_011526413.2 | 1214 | Intron | XP_011524715.1 | |||
XM_011526414.2 | 1214 | Intron | XP_011524716.1 | |||
XM_011526415.2 | 1214 | Intron | XP_011524717.1 |
LENG9 - leukocyte receptor cluster member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301782.1 | 1214 | Silent Mutation | CTA,CTG | L,L 342 | NP_001288711.1 | |
NM_198988.1 | 1214 | Silent Mutation | CTA,CTG | L,L 365 | NP_945339.2 |