Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGCCATTTCTTTGGCATGTCCTT[C/G]AGGAACAGGTCCTGCTGCTCCTCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608791 MIM: 600691 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NXNL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NXNL1 - nucleoredoxin-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138454.1 | 320 | Silent Mutation | CTC,CTG | L,L 91 | NP_612463.1 |
SLC27A1 - solute carrier family 27 member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |