Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTGGGAAATCCATAGCTAAGATG[A/C]CAGGACCCCCTGAAAGCCTAGACAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF493 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF493 - zinc finger protein 493 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001076678.2 | 123 | Intron | NP_001070146.1 | |||
NM_145326.2 | 123 | Missense Mutation | ACA,CCA | T,P 2 | NP_663299.2 | |
NM_175910.6 | 123 | UTR 5 | NP_787106.4 |
ZNF738 - zinc finger protein 738 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |