Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGACTTCATCTTTGCCCACGTG[A/C]GAGGCGAGACCAAGGAGGTGGAGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608792 MIM: 605535 MIM: 188070 | ||||||||||||||||||||
Literature Links: |
GIPC3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GIPC3 - GIPC PDZ domain containing family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133261.2 | 386 | Silent Mutation | AGA,CGA | R,R 107 | NP_573568.1 | |
XM_005259492.3 | 386 | Silent Mutation | AGA,CGA | R,R 107 | XP_005259549.1 |
HMG20B - high mobility group 20B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBXA2R - thromboxane A2 receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |