Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCGGAGCAGCCGAAGATCTTCTT[A/G]CCGCGGCCGGAGCCAGACGGGGGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609133 | ||||||||||||||||||||
Literature Links: |
FIZ1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FIZ1 - FLT3 interacting zinc finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032836.2 | 1320 | Silent Mutation | GGC,GGT | G,G 410 | NP_116225.2 | |
XM_005259352.4 | 1320 | Silent Mutation | GGC,GGT | G,G 410 | XP_005259409.1 | |
XM_011527426.2 | 1320 | Silent Mutation | GGC,GGT | G,G 419 | XP_011525728.1 |
ZNF524 - zinc finger protein 524 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |