Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTAGAGTTGGGACTTTGGACAGG[A/G]CTCTGAAGGTTTGGGAGAGTGAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604638 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACTN4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACTN4 - actinin alpha 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322033.1 | Intron | NP_001308962.1 | ||||
NM_004924.5 | Intron | NP_004915.2 | ||||
XM_005259281.4 | Intron | XP_005259338.1 | ||||
XM_006723406.2 | Intron | XP_006723469.1 | ||||
XM_017027331.1 | Intron | XP_016882820.1 |