Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTCACGGGGTATTGCACTGAGCC[A/G]TCGCGCAACCAGCCCGCGTTGCACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600826 MIM: 606563 | ||||||||||||||||||||
Literature Links: |
HAPLN4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HAPLN4 - hyaluronan and proteoglycan link protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023002.2 | 703 | Silent Mutation | GAC,GAT | D,D 216 | NP_075378.1 |
NCAN - neurocan | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TM6SF2 - transmembrane 6 superfamily member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |