Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAGTTCTGTCACATTCTTGACAT[C/T]TGTAGAGTTTCTCACAAGTATAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF675 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF675 - zinc finger protein 675 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138330.2 | 1486 | Intron | NP_612203.2 | |||
XM_017026442.1 | 1486 | Missense Mutation | AAA,AGA | K,R 153 | XP_016881931.1 | |
XM_017026443.1 | 1486 | Missense Mutation | AAA,AGA | K,R 153 | XP_016881932.1 | |
XM_017026444.1 | 1486 | Intron | XP_016881933.1 |