Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCCGGACACCAGTCCCATCTAAA[C/T]CCCAGGACAAAGTGGAAGCAGCTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605800 | ||||||||||||||||||||
Literature Links: |
CCDC97 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC97 - coiled-coil domain containing 97 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052848.1 | 231 | Missense Mutation | CCC,TCC | P,S 37 | NP_443080.1 | |
XM_005259377.4 | 231 | UTR 5 | XP_005259434.1 | |||
XM_017027442.1 | 231 | Missense Mutation | CCC,TCC | P,S 91 | XP_016882931.1 |
HNRNPUL1 - heterogeneous nuclear ribonucleoprotein U like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |