Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACCTGTCAGAGGAAATGTCACTG[A/C]CAAAGCTGGAGTCGTTCAACGGGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612766 MIM: 611810 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BABAM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BABAM1 - BRISC and BRCA1 A complex member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033549.2 | 525 | Missense Mutation | ACA,CCA | T,P 108 | NP_001028721.1 | |
NM_001288756.1 | 525 | Missense Mutation | ACA,CCA | T,P 108 | NP_001275685.1 | |
NM_001288757.1 | 525 | Missense Mutation | ACA,CCA | T,P 108 | NP_001275686.1 | |
NM_014173.3 | 525 | Missense Mutation | ACA,CCA | T,P 108 | NP_054892.2 |
USHBP1 - USH1 protein network component harmonin binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |