Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATGATATGTAAGTTGTGTAGCAC[A/G]TACAAAGGTCTTCCCACATTCCTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613903 | ||||||||||||||||||||
Literature Links: |
ZNF540 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF540 - zinc finger protein 540 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172225.2 | 2220 | Intron | NP_001165696.1 | |||
NM_001172226.2 | 2220 | Intron | NP_001165697.1 | |||
NM_152606.4 | 2220 | Intron | NP_689819.1 |
ZNF571 - zinc finger protein 571 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290314.1 | 2220 | Missense Mutation | CGT,TGT | R,C 485 | NP_001277243.1 | |
NM_001321272.1 | 2220 | Missense Mutation | CGT,TGT | R,C 485 | NP_001308201.1 | |
NM_016536.4 | 2220 | Missense Mutation | CGT,TGT | R,C 485 | NP_057620.3 | |
XM_011527024.2 | 2220 | Missense Mutation | CGT,TGT | R,C 488 | XP_011525326.1 | |
XM_017026855.1 | 2220 | Missense Mutation | CGT,TGT | R,C 527 | XP_016882344.1 | |
XM_017026856.1 | 2220 | Missense Mutation | CGT,TGT | R,C 488 | XP_016882345.1 | |
XM_017026857.1 | 2220 | Missense Mutation | CGT,TGT | R,C 488 | XP_016882346.1 | |
XM_017026858.1 | 2220 | Missense Mutation | CGT,TGT | R,C 451 | XP_016882347.1 | |
XM_017026859.1 | 2220 | Missense Mutation | CGT,TGT | R,C 451 | XP_016882348.1 | |
XM_017026860.1 | 2220 | Missense Mutation | CGT,TGT | R,C 421 | XP_016882349.1 |
ZNF571-AS1 - ZNF571 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |