Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGGCACTCACAGACAGGCATGTC[A/G]TTGCAGCAGTCCGTGAGACCTGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611764 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNFN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNFN - cornifelin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032488.3 | 154 | Silent Mutation | AAC,AAT | N,N 33 | NP_115877.2 | |
XM_005259332.2 | 154 | Silent Mutation | AAC,AAT | N,N 46 | XP_005259389.1 | |
XM_011527396.1 | 154 | Silent Mutation | AAC,AAT | N,N 46 | XP_011525698.1 | |
XM_011527397.1 | 154 | Silent Mutation | AAC,AAT | N,N 46 | XP_011525699.1 |
LIPE-AS1 - LIPE antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101930071 - uncharacterized LOC101930071 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |