Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGCCGCGAGCACACACCTTCCTC[C/T]CCGTAGCGGTCGAAGATCTCGCGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604572 MIM: 610057 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJB1 - DnaJ heat shock protein family (Hsp40) member B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300914.1 | Intron | NP_001287843.1 | ||||
NM_001313964.1 | Intron | NP_001300893.1 | ||||
NM_006145.2 | Intron | NP_006136.1 | ||||
XM_006722733.1 | Intron | XP_006722796.1 | ||||
XM_006722734.3 | Intron | XP_006722797.1 | ||||
XM_011527956.2 | Intron | XP_011526258.1 |
TECR - trans-2,3-enoyl-CoA reductase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |