Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCATCCTGGCCCAGCTGTACCA[C/T]GAGAAGCCACTGGTGTTCCTGGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605800 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC97 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC97 - coiled-coil domain containing 97 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052848.1 | 425 | Silent Mutation | CAC,CAT | H,H 101 | NP_443080.1 | |
XM_005259377.4 | 425 | Silent Mutation | CAC,CAT | H,H 36 | XP_005259434.1 | |
XM_017027442.1 | 425 | Silent Mutation | CAC,CAT | H,H 155 | XP_016882931.1 |
HNRNPUL1 - heterogeneous nuclear ribonucleoprotein U like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |