Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAGTGCCCAAATGAGCAAGATAT[C/G]GCAGCAAAACAGCACTCCAGGGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 138972 MIM: 613230 | ||||||||||||||||||||
Literature Links: |
CEBPG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CEBPG - CCAAT/enhancer binding protein gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252296.1 | 200 | Missense Mutation | TCG,TGG | S,W 5 | NP_001239225.1 | |
NM_001806.3 | 200 | Missense Mutation | TCG,TGG | S,W 5 | NP_001797.1 |
PEPD - peptidase D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |