Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGCCAGGTGCCTGGCCCGATGTC[A/G]CCGACCCGGAGCCGGTTGTGCGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610055 | ||||||||||||||||||||
Literature Links: |
CC2D1A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CC2D1A - coiled-coil and C2 domain containing 1A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PODNL1 - podocan like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146254.1 | 1512 | Silent Mutation | GGC,GGT | G,G 466 | NP_001139726.1 | |
NM_001146255.1 | 1512 | Silent Mutation | GGC,GGT | G,G 377 | NP_001139727.1 | |
NM_024825.3 | 1512 | Silent Mutation | GGC,GGT | G,G 468 | NP_079101.3 | |
XM_006722901.1 | 1512 | Silent Mutation | GGC,GGT | G,G 468 | XP_006722964.1 | |
XM_006722902.1 | 1512 | Silent Mutation | GGC,GGT | G,G 423 | XP_006722965.1 | |
XM_006722903.1 | 1512 | Silent Mutation | GGC,GGT | G,G 379 | XP_006722966.1 | |
XM_011528308.1 | 1512 | Silent Mutation | GGC,GGT | G,G 466 | XP_011526610.1 | |
XM_011528309.2 | 1512 | Silent Mutation | GGC,GGT | G,G 450 | XP_011526611.1 | |
XM_011528310.2 | 1512 | Silent Mutation | GGC,GGT | G,G 438 | XP_011526612.1 | |
XM_011528311.1 | 1512 | Silent Mutation | GGC,GGT | G,G 435 | XP_011526613.1 | |
XM_011528313.1 | 1512 | Silent Mutation | GGC,GGT | G,G 386 | XP_011526615.1 | |
XM_011528314.1 | 1512 | Silent Mutation | GGC,GGT | G,G 371 | XP_011526616.1 | |
XM_011528316.1 | 1512 | Silent Mutation | GGC,GGT | G,G 297 | XP_011526618.1 | |
XM_017027313.1 | 1512 | Silent Mutation | GGC,GGT | G,G 371 | XP_016882802.1 |