Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCTCCCCACAGGGCTCACCTGCC[C/T]ACGCTTTATCCCAAAGTAGCGCGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601155 MIM: 180664 | ||||||||||||||||||||
Literature Links: |
ARHGAP45 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARHGAP45 - Rho GTPase activating protein 45 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLR2E - RNA polymerase II subunit E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316323.1 | 456 | Missense Mutation | AGG,GGG | R,G 95 | NP_001303252.1 | |
NM_001316324.1 | 456 | Missense Mutation | AGG,GGG | R,G 95 | NP_001303253.1 | |
NM_002695.3 | 456 | Missense Mutation | AGG,GGG | R,G 188 | NP_002686.2 | |
XM_011528070.2 | 456 | Missense Mutation | AGG,GGG | R,G 189 | XP_011526372.1 | |
XM_017026883.1 | 456 | Missense Mutation | AGG,GGG | R,G 96 | XP_016882372.1 |