Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCACCCGCAGGACTTGCACCTTC[A/G]AGATCCCCTGTGTTTACCCAGCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LOC100506634 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC100506634 - hepatocellular carcinoma-associated antigen HCA25a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF584 - zinc finger protein 584 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318002.1 | 657 | Silent Mutation | TCA,TCG | S,S 53 | NP_001304931.1 | |
NM_173548.2 | 657 | Silent Mutation | TCA,TCG | S,S 61 | NP_775819.1 | |
XM_005258628.4 | 657 | Silent Mutation | TCA,TCG | S,S 53 | XP_005258685.1 | |
XM_005258630.4 | 657 | Silent Mutation | TCA,TCG | S,S 61 | XP_005258687.1 |