Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACATCACCTACCTTGGACCCCAC[A/G]GGCTGGAAGCCGGGCAGGCTGCTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614770 MIM: 601717 MIM: 610850 | ||||||||||||||||||||
Literature Links: |
PCP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PCP2 - Purkinje cell protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271830.1 | 288 | Silent Mutation | CCC,CCT | P,P 77 | NP_001258759.1 | |
NM_174895.2 | 288 | Silent Mutation | CCC,CCT | P,P 93 | NP_777555.1 | |
XM_006722639.2 | 288 | Silent Mutation | CCC,CCT | P,P 56 | XP_006722702.1 | |
XM_017026249.1 | 288 | Silent Mutation | CCC,CCT | P,P 209 | XP_016881738.1 | |
XM_017026250.1 | 288 | Silent Mutation | CCC,CCT | P,P 209 | XP_016881739.1 | |
XM_017026251.1 | 288 | Silent Mutation | CCC,CCT | P,P 93 | XP_016881740.1 | |
XM_017026252.1 | 288 | Silent Mutation | CCC,CCT | P,P 56 | XP_016881741.1 |
PET100 - PET100 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171155.1 | 288 | Intron | NP_001164626.1 |
STXBP2 - syntaxin binding protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
XAB2 - XPA binding protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |