Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTCCAAGCGCAGCAGCACCATTT[C/T]GGGGCCCCAGCTGGCCAGCGTGAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609458 | ||||||||||||||||||||
Literature Links: |
MAN2B1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAN2B1 - mannosidase alpha class 2B member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000528.3 | 2386 | Missense Mutation | AAA,GAA | K,E 911 | NP_000519.2 | |
NM_001173498.1 | 2386 | Missense Mutation | AAA,GAA | K,E 910 | NP_001166969.1 | |
XM_005259913.1 | 2386 | Missense Mutation | AAA,GAA | K,E 912 | XP_005259970.1 | |
XM_017026818.1 | 2386 | Missense Mutation | AAA,GAA | K,E 544 | XP_016882307.1 |