Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAGCTGGAACAGATGGAAGACTT[A/C]TTCCTAGATGCCCCGCCCCTCCCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606398 MIM: 609742 MIM: 605815 | ||||||||||||||||||||
Literature Links: |
ATF5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATF5 - activating transcription factor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193646.1 | 722 | Missense Mutation | TTA,TTC | L,F 115 | NP_001180575.1 | |
NM_001290746.1 | 722 | Missense Mutation | TTA,TTC | L,F 115 | NP_001277675.1 | |
NM_012068.5 | 722 | Missense Mutation | TTA,TTC | L,F 115 | NP_036200.2 | |
XM_011526629.2 | 722 | Missense Mutation | TTA,TTC | L,F 115 | XP_011524931.1 |
IL4I1 - interleukin 4 induced 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4751 - microRNA 4751 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUP62 - nucleoporin 62 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |