Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTGTCCCGCAGACGACCGCGGGC[C/G]GCAAAAACCCAGATTACTTGGTTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601901 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MEIS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MEIS3 - Meis homeobox 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC8A2 - solute carrier family 8 member A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015063.2 | 4360 | UTR 3 | NP_055878.1 | |||
XM_005259172.1 | 4360 | UTR 3 | XP_005259229.1 | |||
XM_017027159.1 | 4360 | UTR 3 | XP_016882648.1 |